Learn More
Aviva Systems Biology Corp WASL Antibody - middle region (ARP63498_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63498_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against WASL. It was validated on Western Blot. . Alternate names: DKFZp779G0847, MGC48327, N-WASP, NWASPApplicationWB, IHC. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-287-989