Learn More
WASP BAR CODE Wasp Wireless 2D Scanner, 65 ft. Scan Range, IP65/IP67, Bluetooth 5.0
SDP
Catalog No. NC3983254
Encompass
This ultra-rugged handheld barcode scanner is designed for demanding warehouse and industrial environments. It features an ergonomic design, bright aiming system, and advanced auto range imager to capture 1D and 2D barcodes from inches to 65 ft., even on damaged labels. Connectivity is provided by Bluetooth™ 5.0 with a 328 ft. range. A 3350 mAh battery ensures extended operation. It is built to withstand harsh conditions with an IP65/IP67 seal, -4°F to 122°F operating temperatures, and 6.6 ft. drop resistance.
  • Supports 80,000+ scans per charge for long-lasting use.
  • Ergonomic and balanced design reduces strain during all-day use.
  • Advanced auto range imager provides reliable performance on damaged or shrink-wrapped barcodes.
Catalog No. NC3983254 Supplier WASP BAR CODE Supplier No. I00009450
Add to Cart
Add to Cart
This ultra-rugged handheld barcode scanner is designed for demanding warehouse and industrial environments. It features an ergonomic design, bright aiming system, and advanced auto range imager to capture 1D and 2D barcodes from inches to 65 ft., even on damaged labels. Connectivity is provided by Bluetooth™ 5.0 with a 328 ft. range. A 3350 mAh battery ensures extended operation. It is built to withstand harsh conditions with an IP65/IP67 seal, -4°F to 122°F operating temperatures, and 6.6 ft. drop resistance.
  • Supports 80,000+ scans per charge for long-lasting use.
  • Ergonomic and balanced design reduces strain during all-day use.
  • Advanced auto range imager provides reliable performance on damaged or shrink-wrapped barcodes.