Learn More
Sigma Aldrich Waspaloy - heat resisting alloy, Ni58Cr19Co14MoTiAlFe
Catalog No. 503092332 Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
10 Ea.
Catalog No. Quantity
50-309-2332 10 Ea.
1 options
Catalog No. 50-309-2332 Supplier Sigma Aldrich Supplier No. GF4235306310EA
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Waspaloy - heat resisting alloy, Ni58Cr19Co14MoTiAlFe
Specifications
Quantity 10 Ea.
Synonym NI150300