Learn More
Waters Corp Waters Waste Bottle Trap Assembly, Replacement, Condensate Trap
Catalog No. 50793460 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Waste Bottle Trap Assembly is a replacement component specifically designed to trap condensate in the gas exhaust of compatible Waters analytical instruments.
  • Functions as a replacement part for existing assemblies.
  • Effectively traps condensate from the gas exhaust.
  • Ensures optimal performance and protection of your instrument.
  • Compatible with a wide range of Waters instruments, including ACQUITY TQD, SQD, 3100, ZQ, Quattro Micro API, Quattro Premier, LCT Premier, QTOF Premier, and Synapt.
Catalog No. 50-793-460 Supplier Waters Corp Supplier No. S100970BC2S
This Waste Bottle Trap Assembly is a replacement component specifically designed to trap condensate in the gas exhaust of compatible Waters analytical instruments.
  • Functions as a replacement part for existing assemblies.
  • Effectively traps condensate from the gas exhaust.
  • Ensures optimal performance and protection of your instrument.
  • Compatible with a wide range of Waters instruments, including ACQUITY TQD, SQD, 3100, ZQ, Quattro Micro API, Quattro Premier, LCT Premier, QTOF Premier, and Synapt.