Learn More
California Pacific Lab & Consulting WASTE CONTAINER 5L W/LID RED
SDP
Catalog No. NC9497752
Encompass_Preferred
Bench Top Solid Waste Container With Detachable Base. 5 L Capacity, Prevents 99.9% Emissions Of Toxic Fumes, Hdpe, Red Lid With Gasket That Latches. Prevent Cross Contamination For Small Disposables. Small Footprint, Available With Yellow Lid. Red An
Catalog No. NC9497752 Supplier California Pacific Lab & Consulting Supplier No. SWC002
Bench Top Solid Waste Container With Detachable Base. 5 L Capacity, Prevents 99.9% Emissions Of Toxic Fumes, Hdpe, Red Lid With Gasket That Latches. Prevent Cross Contamination For Small Disposables. Small Footprint, Available With Yellow Lid. Red An