Learn More
HCL Labels Empty - California Compliant Label - 6" x 6" Adhesive Vinyl (Pack of 250)
SDP
Catalog No. NC1977767
Encompass_Preferred
Empty - California Compliant LabelHelpful for marking empty hazardous materials containers, as well as marking empty hazardous waste containers, pails, liners and barrels. This label includes a line for the date and the following verbiage, "This container must be reused, recycled or returned to the supplier within a year of this date. This container is empty by California standards (22 CCR 66261.7(b)). Empty means material can not be scraped or poured from this container."Please note: This item may come in a roll or pack depending on stock status and chosen delivery method. Printed on strong adhesive vinyl.
Catalog No. NC1977767 Supplier HCL Labels Supplier No. CHL0008
Empty - California Compliant LabelHelpful for marking empty hazardous materials containers, as well as marking empty hazardous waste containers, pails, liners and barrels. This label includes a line for the date and the following verbiage, "This container must be reused, recycled or returned to the supplier within a year of this date. This container is empty by California standards (22 CCR 66261.7(b)). Empty means material can not be scraped or poured from this container."Please note: This item may come in a roll or pack depending on stock status and chosen delivery method. Printed on strong adhesive vinyl.