Learn More
DSI LLC WASTE MANIFOLD, 10-Port with SS/Teflon quick disconnect, with (6) 1/4-28, (1) 1/2" NPT, and (3) 1/4" NPT ports, Polypropylene
Catalog No. 501869993
Encompass_Preferred
WASTE MANIFOLD, 10-Port with SS/Teflon quick disconnect, with (6) 1/4-28, (1) 1/2" NPT, and (3) 1/4" NPT ports, Polypropylene
Catalog No. 50-186-9993 Supplier DSI LLC Supplier No. MA1003
WASTE MANIFOLD, 10-Port with SS/Teflon quick disconnect, with (6) 1/4-28, (1) 1/2" NPT, and (3) 1/4" NPT ports, Polypropylene