Learn More
Cole-Parmer Cole-Parmer VapLock™ Solvent Waste Manifold, PP, 70mm Rieke, 1x1/2"NPT, 3x1/4"NPT, 6x1/4-28UNF; 1/EA
Catalog No. NC1503368 Shop All Cole-Parmer Products
Encompass_Preferred
Select from various manifold cap sizes to fit your laboratory and industrial solvent waste containers. Jars and pail caps are available with polypropylene (PP) or PTFE manifolds and 4 or 10 variously sized ports. Ports accept male thread adapters to connect flexible and rigid tubing, or filter 12019-79 or filter/indicator 12019-80.
Catalog No. NC1503368 Supplier Cole-Parmer Supplier No. 1201924
Select from various manifold cap sizes to fit your laboratory and industrial solvent waste containers. Jars and pail caps are available with polypropylene (PP) or PTFE manifolds and 4 or 10 variously sized ports. Ports accept male thread adapters to connect flexible and rigid tubing, or filter 12019-79 or filter/indicator 12019-80.