Learn More
BD Cell Analysis 3P BD Biosciences Waste Sensor Probe, 2-Level Sensor, 10.6 in Stem Length, 120 VAC
Catalog No. NC0244759
Encompass
The BD Biosciences Waste Sensor Probe is designed for monitoring waste fluid levels in scientific instrumentation, enhancing operational safety and reliability. It features a 10.6 in stem, operates at 120 VAC, and provides a 2-level sensing mechanism. Manufactured under strict GMP guidelines for Research Use Only (RUO), this product ensures compliance with high manufacturing standards while offering a reliable solution for waste level monitoring in laboratory settings.
Catalog No. NC0244759 Supplier BD Cell Analysis 3P Supplier No. 338978
Add to Cart
Add to Cart
The BD Biosciences Waste Sensor Probe is designed for monitoring waste fluid levels in scientific instrumentation, enhancing operational safety and reliability. It features a 10.6 in stem, operates at 120 VAC, and provides a 2-level sensing mechanism. Manufactured under strict GMP guidelines for Research Use Only (RUO), this product ensures compliance with high manufacturing standards while offering a reliable solution for waste level monitoring in laboratory settings.