Learn More
Sigma Aldrich Fine Chemicals Biosciences Waste solvent funnels for1EA
Catalog No. 502901366
Encompass
ECO funnels prevent waste solvent evaporation. We recommend that the ECO funnel be removed from the waste container at the end of each day or when it is not in use for long periods of time.SpecificationsChemically resistant HDPE and PP constructionLip-seal on lid (red) seals the waste system when closedfunnel stem sealed to waste container cap prevents emission from sides of stemPressure-equalizing tube prevents solvent trappingfunnels are sized to fit the following waste solvent containers4 L funnel fits round bottle B84108 L funnel fits round bottle B85359 L funnel fits rectangular carboy C631720 L funnel fits rectangular D.O.T. carboy Z268208Order funnel and waste container separately.
Catalog No. 50-290-1366 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z2865161EA
ECO funnels prevent waste solvent evaporation. We recommend that the ECO funnel be removed from the waste container at the end of each day or when it is not in use for long periods of time.SpecificationsChemically resistant HDPE and PP constructionLip-seal on lid (red) seals the waste system when closedfunnel stem sealed to waste container cap prevents emission from sides of stemPressure-equalizing tube prevents solvent trappingfunnels are sized to fit the following waste solvent containers4 L funnel fits round bottle B84108 L funnel fits round bottle B85359 L funnel fits rectangular carboy C631720 L funnel fits rectangular D.O.T. carboy Z268208Order funnel and waste container separately.