Learn More
ERA WATERS CORP WASTEWATR COLIFORMS QC
SDP
Catalog No. NC0645115
Encompass
WasteWatR Coliforms Two lyophilized samples consist of one negative sample and one positive to be quantitated for total, fecal and E. coli coliforms.
Catalog No. NC0645115 Supplier ERA WATERS CORP Supplier No. 083
WasteWatR Coliforms Two lyophilized samples consist of one negative sample and one positive to be quantitated for total, fecal and E. coli coliforms.