Learn More
Cole-Parmer Watch Glass 25mm 12/pk
Supplier:  Cole-Parmer 3450846
Encompass_Preferred
Cole-Parmer Watch Glass 25mm 12/pk, Manufactured from clear glass sheet (1.8 to 2.0mm thick), these watch glasses are fire ground and have polished edges. Concave shape prevents sliding and return of condensation to the beaker.
Catalog No. NC1217916