Learn More
Cole-Parmer Essentials Watch Glass, 65 mm, fits 150 mL beakers, 12/pk
Supplier:  Cole-Parmer 3450852
Encompass_Preferred
Manufactured from clear glass sheet (1.8 to 2.0mm thick), these watch glasses are fire ground and have polished edges. Concave shape prevents sliding and return of condensation to the beaker.
Catalog No. NC1889317