Learn More
Foxx Life Sciences Borosil® High Clarity S-Line Watch Glass with Flat Curved Surface, 120mL, 20/CS
Supplier:  Foxx Life Sciences 9986079
Encompass_Preferred
Borosil® Watch Glasses (S-Line) are made of soda glass & each is molded to the same radius of curvature for convenient stacking. The watch glasses are high clarity & free of bubbles. A special process is used that results in an ideal flat curved surface. Avail. in capacity of 80ml to 150ml. Watch glasses have many usages from being a surface to evaporate a liquid on, to holding solids while being weighed, for heating small amounts of substances. In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil® Type 3.3 Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil® Laboratory Glassware offers unparalleled mechanical strength, clarity, & reliability at competitive prices. & thanks to a unique annealing process, Borosil® Glassware is stress-free & incredibly resistant to thermal shock.
Catalog No. 50-172-0446