Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety Hi-Vis Orange Nylon Work Gloves with Foam Nitrile Coated Palm

Catalog No. 19812710 Shop All MCR Safety Products
Encompass_Preferred
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-812-710 2X-Large
19-812-708 Small
19-812-707 Medium
19-812-706 Large
19-812-709 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-812-710 Supplier MCR Safety Supplier No. 96790HVIXXL
May include imposed supplier surcharges.
Add to Cart
Add to Cart
This item is not returnable. View return policy

A printed safety reminder on the back of each glove reminds users to watch their hands at all times. Hi-vis orange nitrile foam on the palm provides abrasion resistance and enhances wet/dry grip capabilities.

  • Nylon shell is comfortable and flexible for optimal dexterity
  • Hi-vis orange BNF (Breathable Nitrile Foam) with NFT™ coating on the palm and fingertips
  • Nitrile foam provides abrasion resistance and enhanced grip capabilities
  • Watch Your Hands logo printed on the back of each glove as a constant safety reminder
  • ActiFresh™ treatment reduces bacteria and odors

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance ANSI/ISEA 105 Cut Level A2, ANSI/ISEA 105 Abrasion Level 4, ANSI/ISEA 105 Puncture Level 2, EN 388:2016 4141, OEKO-TEX STANDARD 100
Certifications/Compliance (Europe) EN 388:2016 4141, OEKO-TEX™ STANDARD 100
Construction Seamless Knit
Coating Material Nitrile Foam
Color Hi-vis Orange, Gray
Exterior Finish No Texture
Grade Premium
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size 2X-Large
Finger Style Full
For Use With (Application) Hand Protection
Gender Unisex
Glove Cuff Style Knit
Grip Design Textured
Puncture Rating ANSI/ISEA 105-16 Level 2
Series 96790HVI
Yarn Gauge 15
Product Line Watch Your Hands
Show More Show Less

WARNING: Cancer and Reproductive Harm www.P65Warnings.ca.gov/product

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.