Learn More
MSC Majestic Glove Cut-Less Watchdog Glove, Foam Nitrile, Cut A4, Extra Small, 1 ea
Catalog No. 5000688416
Encompass
These 18-gauge seamless knit gloves provide exceptional dexterity and superior cut protection for demanding industrial applications. Engineered with Korplex® technology, the gloves feature a pre-curved finger design to minimize hand fatigue and offer a comfortable, secure fit during extended use.
  • Seamless knit construction with 18-gauge Korplex® cut-resistant technology
  • Premium breathable foam nitrile palm coating for superior grip, durability, and dexterity
  • Touch screen capable for use with most smartphones and tablets
  • Sanitized® by Actifresh coating to inhibit bacteria and minimize odor
  • Reinforced thumb crotch for added durability
  • Long knit cuff for a secure fit
  • Pre-curved finger design to reduce hand fatigue
  • Latex free and crafted without irritant fibers for comfort
  • Meets ANSI/ISEA 105-2024 cut level A4 and EN 388:2016 4X42D standards
Catalog No. 50-006-88416 Supplier MSC Supplier No. 99464968
These 18-gauge seamless knit gloves provide exceptional dexterity and superior cut protection for demanding industrial applications. Engineered with Korplex® technology, the gloves feature a pre-curved finger design to minimize hand fatigue and offer a comfortable, secure fit during extended use.
  • Seamless knit construction with 18-gauge Korplex® cut-resistant technology
  • Premium breathable foam nitrile palm coating for superior grip, durability, and dexterity
  • Touch screen capable for use with most smartphones and tablets
  • Sanitized® by Actifresh coating to inhibit bacteria and minimize odor
  • Reinforced thumb crotch for added durability
  • Long knit cuff for a secure fit
  • Pre-curved finger design to reduce hand fatigue
  • Latex free and crafted without irritant fibers for comfort
  • Meets ANSI/ISEA 105-2024 cut level A4 and EN 388:2016 4X42D standards