Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water with 0.1% Trifluoroacetic acid, LabReady™ Solvent Blend, Solstice

Catalog No. 6002516 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
4 x 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-025-16 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-025-16 Supplier Solstice Supplier No. 4854
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7732-18-5 , 76-05-1
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide

Specifications

Boiling Point 100°C
CAS Max % 99.9000%, 0.1000%
Density 0.9982 g/cm3
Grade Solvent Blend
Packaging Glass Bottle
Quantity 4 x 4 L
Chemical Name or Material Water with 0.1% Trifluoroacetic acid, LabReady™ Solvent Blend
ShelfLife 365 days from date of manufacture
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.