Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water with 0.1% (v/v) Acetic Acid, Hypergrade, For LC-MS LiChrosolv™, MilliporeSigma™

Catalog No. 01100256 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-100-256 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-100-256 Supplier MilliporeSigma Supplier No. 1.59007.2500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Boiling Point 100°C (1013 hPa)
Grade Hypergrade
Packaging Glass Bottle
Quantity 2.5 L
Recommended Storage 2 to 30 °C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.