Learn More
Medchemexpress LLC Water-17O | 13768-40-6 | 99.0% | 19.02 | 10 MG
SDP
Catalog No. 5000345815
Encompass_Preferred
Water-17O is the 17O labeled Water. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotope
Catalog No. 50-003-45815 Supplier Medchemexpress LLC Supplier No. HY108872S11MG
Water-17O is the 17O labeled Water. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotope