Learn More
Medchemexpress LLC Water-17O | 13768-40-6 | 99.0% | 19.02 g/mol | H2(17)O | 5 MG
SDP
Catalog No. 5000264652
Encompass_Preferred
Water-17O is water isotopically enriched with the stable oxygen-17 (17O) isotope for laboratory and analytical applications. It is intended for research use as a tracer and as an internal standard in quantitative spectroscopic and mass-spectrometry methods.
  • Isotopically labeled with oxygen-17 for tracer and standard applications.
  • High isotopic purity: 99.0%.
  • Compatible with NMR, GC-MS, and LC-MS workflows.
  • Useful for quantitative analysis and method development.
Catalog No. 50-002-64652 Supplier Medchemexpress LLC Supplier No. HY108872S15MG
Water-17O is water isotopically enriched with the stable oxygen-17 (17O) isotope for laboratory and analytical applications. It is intended for research use as a tracer and as an internal standard in quantitative spectroscopic and mass-spectrometry methods.
  • Isotopically labeled with oxygen-17 for tracer and standard applications.
  • High isotopic purity: 99.0%.
  • Compatible with NMR, GC-MS, and LC-MS workflows.
  • Useful for quantitative analysis and method development.