Learn More
Medchemexpress LLC Water-18O | 14314-42-2 | 99.99% | 20.02 | 250 MG
SDP
Catalog No. 5000347721
Encompass_Preferred
Water-18O is the 18O-labeled Water. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. For research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-47721 Supplier Medchemexpress LLC Supplier No. 250MGHY108872S
Water-18O is the 18O-labeled Water. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. For research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.