Learn More
Thermo Scientific™ Water Analysis Instrument Calibration and Repair Service
Catalog No. 15200961
Change view
Click to view available options
Product Type:
Calibration Level 1
Calibration Level 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
15-200-961 Calibration Level 1
15-200-962 Calibration Level 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 15-200-961 Supplier Thermo Scientific™ Supplier No. CALSERVD1
Add to Cart
Add to Cart
Thermo Scientific™ provides calibration and repair services for electrochemistry devices for water analysis including pH meters. Instruments are issued calibration certificates that are traceable to NIST standards and can be used for internal compliance and audit.
Out of tolerance instruments can be adjusted or repaired as needed. Thermo Scientific™ also offers cost-effective device exchanges and loaner services, as well as expedited services for devices that need to be repaired within 24 - 48 hours.
Benefits:
  • Helps meet ISO, FDA, USDA, EPA, GLP/cGMP quality assurance guidelines before use in critical processes
  • Instruments are calibrated to factory standards and include before (as found) and after (as left) data to
    document instrument accuracy
  • Gain confidence in measurement precision
  • Traceability chain for the standards used in calibration
  • Protect your investment and prevent equipment downtime
  • pH calibration service includes measurement of meter accuracy at five test points across pH range and five test points across corresponding mV range
  • Calibration services performed by highly skilled technicians and backed by a 90-day satisfaction guarantee
To help meet regulatory requirements, all Thermo Scientific™ Orion™ Water Analysis Instrument Test Equipment is traceable to NIST.
Certification of Thermo Scientific™'s test equipment is compliant with ANSI/NCSL ZA540-1 standards, ISO 9001:2015, and ISO 9001:2005 requirements.
Annual calibrations from the date your equipment is put into operation are recommended.
Thermo Scientific™'s calibration services specialize in servicing the following Thermo Scientific™ Orion™ Water Analysis Instrumentation:
  • Thermo Scientific™ Orion™ Versa Star Pro™
  • Thermo Scientific™ Orion™ Dual Star™
  • Thermo Scientific™ Orion Star A™
  • Thermo Scientific™ Orion Star™
  • Thermo Scientific™ Orion Star™ T900 Series
  • Thermo Scientific™ Orion™ Conductivity Resistor Kits
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Orion Star A111, Star A112, Star A113, Star A121, Star A122, Star A123, Star A211, Star A212, Star A213, Star A214, Star A221, Star A222, Star A223, Star A321, Star A322, Star A323, Star A324, 0261S0, 0266S0, 0131S3, 0136S3, Versa Star (1 Module), 2-Star/2-Star Plus, 3-Star/3-Star Plus; Accumet: AB150, AB300, AB315, XL150, AP71, AP72, AP74, AP75, AP110, AP115
Product Type Calibration Level 1
Product Line Thermo Scientific
Service Offering Calibration and Repair
For Use With (Application) Electrochemistry devices for water analysis including pH meters