Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water, for HPLC and Spectrophotometry, Solstice

Catalog No. 6003105 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
20 L
Packaging:
Hdpe Nowpak™
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-031-05 20 L Hdpe Nowpak™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-031-05 Supplier Solstice Supplier No. NP36520
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Boiling Point 100°C
CAS Max % 100.0000%
Beilstein 2050024
Formula Weight 18
Grade HPLC
Packaging Hdpe Nowpak™
Density 0.9971 g/cm3
Quantity 20 L
Chemical Name or Material Water, B&J Brand™, for HPLC and Spectrophotometry
ShelfLife 547 days from date of manufacture
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.