Learn More
Polyscience 20L Digital Water Bath (Ambient +5 to 100°C); 120V, 60Hz
SDP
Catalog No. NC1640537
Encompass_Preferred
PolyScience™ 20 Liter Digital Water BathAmbient +5 to 99°C Working Temperature Range120V, 60HzWorking Access (L x W x D):9.5 x 17 x 6 in24.1 x 43.2 x 15.2 cmTemperature Stability:±0.1°CIncluded Hardware: Sample TrayFlammability Class (DIN 12876-1):I (NFL)Features:• Operates in 5 languages• Displays actual and set point temperature simultaneously• 5 Programmable pre-sets for frequently used temperatures• Integrated timer• Hinged gable cover• Calibration capability• Reservoir made of 304 Stainless Steel• Reservoir drain (10 liter and larger baths)• Primary and automatic safety thermostats• Compatible with waterless bath beads
Catalog No. NC1640537 Supplier Polyscience Supplier No. WBE20A11B
PolyScience™ 20 Liter Digital Water BathAmbient +5 to 99°C Working Temperature Range120V, 60HzWorking Access (L x W x D):9.5 x 17 x 6 in24.1 x 43.2 x 15.2 cmTemperature Stability:±0.1°CIncluded Hardware: Sample TrayFlammability Class (DIN 12876-1):I (NFL)Features:• Operates in 5 languages• Displays actual and set point temperature simultaneously• 5 Programmable pre-sets for frequently used temperatures• Integrated timer• Hinged gable cover• Calibration capability• Reservoir made of 304 Stainless Steel• Reservoir drain (10 liter and larger baths)• Primary and automatic safety thermostats• Compatible with waterless bath beads