Learn More
Azzota Corp Water bath 22L LED
Catalog No. 5000887287
Encompass_Preferred
SINGLE CHAMBER WATER BATH RT 5 to 100C 22L 20kg 450x300x160mm 220V/50Hz LED display
Catalog No. 50-008-87287 Supplier Azzota Corp Supplier No. SDK22
SINGLE CHAMBER WATER BATH RT 5 to 100C 22L 20kg 450x300x160mm 220V/50Hz LED display