Learn More
Polyscience 2L Digital Water Bath (Ambient +5 to 100°C); 120V, 60Hz
SDP
Catalog No. NC1740114
Encompass_Preferred
PolyScience™ 2 Liter Digital Water BathAmbient +5 to 99°C120V, 60HzWorking Access (L x W x D):3.9 x 4.3 x 6 in 9.9 x 10.9 x 15.2 cmTemperature Stability:±0.1°CIncluded Hardware: Sample TrayFlammability Class (DIN 12876-1):I (NFL)Features:• Operates in 5 languages• Displays actual and set point temperature simultaneously• 5 Programmable pre-sets for frequently used temperatures• Integrated timer• Hinged gable cover• Calibration capability• Reservoir made of 304 Stainless Steel• Primary and automatic safety thermostats• Compatible with waterless bath beads
Catalog No. NC1740114 Supplier Polyscience Supplier No. WBE02A11B
PolyScience™ 2 Liter Digital Water BathAmbient +5 to 99°C120V, 60HzWorking Access (L x W x D):3.9 x 4.3 x 6 in 9.9 x 10.9 x 15.2 cmTemperature Stability:±0.1°CIncluded Hardware: Sample TrayFlammability Class (DIN 12876-1):I (NFL)Features:• Operates in 5 languages• Displays actual and set point temperature simultaneously• 5 Programmable pre-sets for frequently used temperatures• Integrated timer• Hinged gable cover• Calibration capability• Reservoir made of 304 Stainless Steel• Primary and automatic safety thermostats• Compatible with waterless bath beads