Learn More
Azzota Corp Water bath 30L LED
Catalog No. 5000887309
Encompass_Preferred
SINGLE CHAMBER WATER BATH RT 5 to 100C 30L 20kg 760x360x325mm 220V/50Hz LED display
Catalog No. 50-008-87309 Supplier Azzota Corp Supplier No. SDK30
SINGLE CHAMBER WATER BATH RT 5 to 100C 30L 20kg 760x360x325mm 220V/50Hz LED display