Learn More
Grainger Water Bath 4L 20.32 x15.79 x32.2
Catalog No. 502752657
Encompass_Preferred
The Benchmark Scientific 4L Water Bath is a general purpose laboratory water bath with a 4 liter capacity stainless steel tank. It has a temperature range of ambient +5 to 99.9°C, stability of ±0.1°C, and uniformity of ±0.5°C. This water bath has over-temperature protection, low liquid level protection, and a front panel digital PID controller. It measures 11.4 x 10.2 x 6.3 inches (WxDxH) and weighs 7.7 lbs. This 120V water bath is CE & RoHS compliant.
Catalog No. 50-275-2657 Supplier Grainger Supplier No. 14N147
The Benchmark Scientific 4L Water Bath is a general purpose laboratory water bath with a 4 liter capacity stainless steel tank. It has a temperature range of ambient +5 to 99.9°C, stability of ±0.1°C, and uniformity of ±0.5°C. This water bath has over-temperature protection, low liquid level protection, and a front panel digital PID controller. It measures 11.4 x 10.2 x 6.3 inches (WxDxH) and weighs 7.7 lbs. This 120V water bath is CE & RoHS compliant.