Learn More
Electron Microscopy Sciences Tissue Grip™ Water Bath Adhesive 1 GL
SDP
Catalog No. 501928644
Encompass_Preferred
Liquid water bath adhesive used in H & E staining, special stains, and immunoperoxidase procedures.
Catalog No. 50-192-8644 Supplier Electron Microscopy Sciences Supplier No. 7130302
Liquid water bath adhesive used in H & E staining, special stains, and immunoperoxidase procedures.