Learn More
Thomas Scientific General Purpose Water Bath, 6 L
SDP
Catalog No. 501458351
Encompass_Preferred
Accurate Temperature Monitoring Labnet's user friendly 6 Liter Water Bath is solidly constructed with a stainless steel, seamless chamber that resists corrosion, a stainless gable cover is included to minimize evaporation, and offers simplicity, consitiancy, and conveneince. The optional non-toxic (non-mercury) thermometer clips onto the gable cover for accurate temperature monitoring. Compact Design Stainless steel gable cover Stainless steel, seamless chamber 550W heating power Pilot light to indicate heating
Catalog No. 50-145-8351 Supplier Thomas Scientific Supplier No. 1195V44
Accurate Temperature Monitoring Labnet's user friendly 6 Liter Water Bath is solidly constructed with a stainless steel, seamless chamber that resists corrosion, a stainless gable cover is included to minimize evaporation, and offers simplicity, consitiancy, and conveneince. The optional non-toxic (non-mercury) thermometer clips onto the gable cover for accurate temperature monitoring. Compact Design Stainless steel gable cover Stainless steel, seamless chamber 550W heating power Pilot light to indicate heating