Learn More
Innovive Inc Mouse Water Bottle - Amber tint - 300ml
Catalog No. NC1966701
Encompass
This item has a minimum order quantity of 200 per supplier requirements. Water Bottle, Amber Color. Amber-colored empty mouse water bottle for use with light-sensitive medications. All bottles are delivered in a bag and irradiated.
Catalog No. NC1966701 Supplier Innovive Inc Supplier No. MWBAMBER
This item has a minimum order quantity of 200 per supplier requirements. Water Bottle, Amber Color. Amber-colored empty mouse water bottle for use with light-sensitive medications. All bottles are delivered in a bag and irradiated.