Learn More
Growcells WATER CELL CULTURE 10L EA
Catalog No. 501034772
Encompass_Preferred
Certified cell culture water, <0.005 EU/mL, double distilled, 0.1um sterile filtered and steam sterilized in TriForest polycarbonate bottles.
Catalog No. 50-103-4772 Supplier Growcells Supplier No. CCPW010L
Certified cell culture water, <0.005 EU/mL, double distilled, 0.1um sterile filtered and steam sterilized in TriForest polycarbonate bottles.