Learn More
Growcells WATER CELL CULTURE 2L EA
Catalog No. 501034770
Encompass_Preferred
Certified cell culture water, <0.005 EU/mL, double distilled, 0.1um sterile filtered and steam sterilized in TriForest polycarbonate bottles.
Catalog No. 50-103-4770 Supplier Growcells Supplier No. CCPW2000
Certified cell culture water, <0.005 EU/mL, double distilled, 0.1um sterile filtered and steam sterilized in TriForest polycarbonate bottles.