Learn More
Growcells WATER CELL CULTURE 2L PK6
Catalog No. 501034771
Encompass_Preferred
Certified cell culture water, <0.005 EU/mL, double distilled, 0.1um sterile filtered and steam sterilized in TriForest polycarbonate bottles.
Catalog No. 50-103-4771 Supplier Growcells Supplier No. CCPW20006
Certified cell culture water, <0.005 EU/mL, double distilled, 0.1um sterile filtered and steam sterilized in TriForest polycarbonate bottles.