Learn More
AVANTOR MASTERFLEX (VWR LLC) Hand-operated chemical siphon/drum pump, 12 strokes/gallon
Catalog No. NC9838949
Encompass
Ideal for most acid/base or ionic salt applications. Portable and self-priming. Includes 60-inch outlet hosecut to fit your application.
Catalog No. NC9838949 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX7060712
Add to Cart
Add to Cart
Ideal for most acid/base or ionic salt applications. Portable and self-priming. Includes 60-inch outlet hosecut to fit your application.