Learn More
Gentech Scientific Inc Recirculating Water Chiller (H150-1000)
SDP
Catalog No. 501897503
Encompass_Preferred
• Temperature Range: 8 - 35 • Temperature Stability: ±0,1 • Temperature Control Mode: PID • Cooling Capacity: 1000 Watt • Pump: Flux-6 l/min at 60 psi, Power-135 Watts • 1 Year Warranty GenTech Scientific offers installation, and extended warranty options. Please contact 585-492-1068 with questions.
Catalog No. 50-189-7503 Supplier Gentech Scientific Inc Supplier No. GTS10972
Add to Cart
Add to Cart
Request a Quote
• Temperature Range: 8 - 35 • Temperature Stability: ±0,1 • Temperature Control Mode: PID • Cooling Capacity: 1000 Watt • Pump: Flux-6 l/min at 60 psi, Power-135 Watts • 1 Year Warranty GenTech Scientific offers installation, and extended warranty options. Please contact 585-492-1068 with questions.