Learn More
Gentech Scientific Inc Recirculating Water Chiller (H150-3000)
SDP
Supplier:  Gentech Scientific Inc GTS10973
Encompass_Preferred
• Temp Range: 8°C - 35°C • Cooling Capacity: 1000 Watt • Pump: Flux-6 l/min at 60 psi, Power-135 Watts • 1 Year Warranty GenTech Scientific offers installation, and extended warranty options. Please contact 585-492-1068 with questions.
Catalog No. 50-189-7504
Add to Cart
Request a Quote