Learn More
Ethos Biosciences Deionized Water, 16oz, Ethos
Catalog No. 5000574211
Encompass_Preferred
Deionized (DI) Water is a water solution suitable for clinical, research and general laboratory applications. Manufactured to meet CLSI reagent grade water standards, it is ideal for slide preparation, reagent formulation, staining procedures, and instrument rinsing. This clear, particle-free solution provides consistent quality for reliable diagnostic and analytical results. It is also widely used for buffer preparation, dilution, cleaning, and other routine laboratory tasks. Supplied in chemically compatible containers, DI Water is appropriate for clinical, research, pharmaceutical, and industrial laboratories requiring dependable reagent grade water.
Catalog No. 50-005-74211 Supplier Ethos Biosciences Supplier No. 331516
Deionized (DI) Water is a water solution suitable for clinical, research and general laboratory applications. Manufactured to meet CLSI reagent grade water standards, it is ideal for slide preparation, reagent formulation, staining procedures, and instrument rinsing. This clear, particle-free solution provides consistent quality for reliable diagnostic and analytical results. It is also widely used for buffer preparation, dilution, cleaning, and other routine laboratory tasks. Supplied in chemically compatible containers, DI Water is appropriate for clinical, research, pharmaceutical, and industrial laboratories requiring dependable reagent grade water.