Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Water-Cooled Condensers for -86°C Upright and Chest Freezers

Catalog No. 11676215 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
12.7, 17, and 20 cu. ft. -86°C Chest Freezers
13.0 cu. ft. -86°C Upright Freezers
17.3, 23, and 28 cu. ft. -86°C Upright Freezers
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
11676215 12.7, 17, and 20 cu. ft. -86°C Chest Freezers
11676213 13.0 cu. ft. -86°C Upright Freezers
11676214 17.3, 23, and 28 cu. ft. -86°C Upright Freezers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11676215 Supplier Thermo Scientific™ Supplier No. 195967
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Work in a cooler environment with the Thermo Scientific™ Water-Cooled Condensers for −86°C upright and chest freezers, which emit 50% less heat into the lab.

Work in a cooler environment with the Thermo Scientific™ Water-Cooled Condensers for -86°C upright and chest freezers, which emit 50% less heat into the lab compared to other freezers.

Features:
  • Work environment stays cooler because 50% less heat is emitted to surroundings (i.e. less BTU rejection).
  • Water regulating valve automatically controls the water flow needed to maintain the compressor discharge pressure.
  • Cooler compressor temperatures enhance compressor stability and reliability.

Requires:

Installation requires a qualified technician and should include proper adjustment of the regulating valve, which controls the discharge pressure.

Note:

Actual requirements for tower and city water sources may vary due to specific water pressure conditions.

TRUSTED_SUSTAINABILITY

Specifications

Type Water-Cooled Condenser
Capacity (English) 20 cu. ft.
Capacity (Metric) 566.3 L
Temperature Range -86°C
Certifications/Compliance CE, UL listed to U.S. and Canadian requirements
Style Upright
Condenser Type Water Cooled
For Use With (Equipment) 12.7, 17, and 20 cu. ft. -86°C Chest Freezers
Defrost Automatic
Compressor Water-cooled
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.