Learn More
System Biosciences LLC Water-soluble Cumate Solution (10,000x)Water-soluble Cumate solution (300mg/ml, 10,000x) = 500 µL
SDP
Catalog No. NC1061274
Encompass_Preferred
Control the cumate-inducible promoter used in SBI’s SparQ Lentivectors with this 10,000x water-soluble inducer solution.With SBI’s SparQ™ Cumate Switch System, you can get inducible gene expression in mammalian cells through the addition of cumate, a non-toxic small molecule. Expression levels are tightly controlled and increase with increasing cumate concentration until maximum induction is reached—see as much as a 32-fold increase in gene expression. Even better, induction is reversible, so you can turn expression levels on and off. Delivering negligible background expression in the absence of cumate, the SparQ System is an excellent choice for achieving controlled levels of gene expression.
Catalog No. NC1061274 Supplier System Biosciences LLC Supplier No. QM150A1
Control the cumate-inducible promoter used in SBI’s SparQ Lentivectors with this 10,000x water-soluble inducer solution.With SBI’s SparQ™ Cumate Switch System, you can get inducible gene expression in mammalian cells through the addition of cumate, a non-toxic small molecule. Expression levels are tightly controlled and increase with increasing cumate concentration until maximum induction is reached—see as much as a 32-fold increase in gene expression. Even better, induction is reversible, so you can turn expression levels on and off. Delivering negligible background expression in the absence of cumate, the SparQ System is an excellent choice for achieving controlled levels of gene expression.