Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Water Cycle Model
GREENER_CHOICESCI_ED

Catalog No. S81680 Shop All Supertek Scientific Products
Change view
Click to view available options
Grade:
3+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S81680 3+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S81680 Supplier Supertek™ Supplier No. B19055
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This 3D Water Cycle Model will show upper level of an underground surface in which soil or rock is permanently saturated with water

  • 3D Water Cycle Model
  • Shows upper level of an underground surface in which soil or rock is permanently saturated with water
  • The water table separates the underground water zone from the capillary fringe or zone of aeration

Specifications

Grade 3+
Product Type Teaching Model
Includes 3D Model
For Use With (Application) Teaching The Water Cycle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.