Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Water Cycle Model in Action
GREENER_CHOICESCI_ED

Catalog No. S89526 Shop All Supertek Scientific Products
Change view
Click to view available options
Grade:
3+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S89526 3+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S89526 Supplier Supertek™ Supplier No. B19052SP
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Make it rain in your classroom. Students can duplicate the individual processes of the water cycle (evaporation condensation and precipitation) with this ingenious model.

  • This Hands-on Model will surely keep kids interested in learning about Evaporation, Condensation and Precipitation.
  • Parts included in this product:
    • Vacuum formed model with cover - 17 x 12.5 x 5 in.
    • Wire Stand
    • Cloud (Ice holder)
    • 10 - 8 x 11 in. Full color Study Cards
    • Companion Guide
  • Requires a heat lamp (not included)
  • Ages 9+

Specifications

Grade 3+
For Use With (Equipment) Heat Lamp
Product Type Supertek™ Water Cycle Model
Includes Vacuum Formed Model with Cover, Wire Stand, Cloud (Ice Holder), Full Color Study Cards, Companion Guide
For Use With (Application) Demonstrate the Water Cycle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.