Learn More
Hach Lange™ Water, deionised and demineralised
Catalog No. 501994227 Shop All Hach Company Products
Encompass
Change view
Click to view available options
Volume (Metric):
4 L
Catalog No. Volume (Metric)
50-199-4227 4 L
1 options
Catalog No. 50-199-4227 Supplier Hach Lange™ Supplier No. 27256
Demineralised Type 2 Water.
Demineralised Type 2 WaterSpecific Conductance: ≤1 μS/cm,Silica (SiO2): ≤30 μg/L,Chloride (Cl-): ≤20 μg/L,Hardness: Negative,Consumption of KMnO4: Negative
Specifications
Platform: Solution
Quantity: 4 L
Shelf Life: 60 months from production date
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 4 L
Type Water Testing Kit Supplies and Reagents