Learn More
Hach Company Water-Deionized-1000 mL
Supplier:  Hach Company 27253
Encompass
Demineralized Type 2 water is used for various laboratory applications that require high ionic purity It is commonly used for preparing solutions and buffers feeding clinical analyzers and performing general chemistry tasks While Type 2 water is not sterile it is ideal for situations where maximum purity is not necessary but higher ionic quality than basic tap water is required Demineralized Type 2 water is used for a wide range of general laboratory work including sample and standard dilution preparation of buffers and dilute chemical reagents and for preparing media to grow bacteria and other microorganisms
Catalog No. 50-002-56968