Learn More
Hach Company Water, Deionized, 100 mL
Supplier:  Hach Company 27242
Encompass
(Demineralized Type 2 Water) Specific Conductance: ≤1 μS/cm, Silica (SiO2): ≤30 μg/L, Chloride (Cl-): ≤20 μg/L, Hardness: Negative, Consumption of KMnO4: Negative
Catalog No. 50-199-4224