Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water, Deionized, Vion Biosciences
SCI_ED

Catalog No. S02513 Shop All Aldon Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S02513 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S02513 Supplier Aldon Corporation Supplier No. VNDD00121L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-quality deionized water, excellent for histology applications.

  • High-quality deionized water, excellent for histology applications.

Chemical Identifiers

Molecular Formula H2O
Molecular Weight (g/mol) 18.01 g/mol
MDL Number MFCD00011332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Hydrogen Oxide, Distilled Water
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name Oxidane
SMILES O

Specifications

pH Data not available.
Color Colorless
Melting Point 0°C
Boiling Point 100°C
CAS Max % 100%
CAS 7732-18-5
CAS Min % 100%
Conductivity 0.0055 microsiemens
Description Deionized Water
Assay Percent Range 100%
Linear Formula HOH
Solubility Information Complete
Formula Weight 18.01 g/mol
Percent Purity 100%
Grade Histological
Concentration or Composition (by Analyte or Components) Pure
Odor No odor.
Packaging Natural Plastic Cylinder
Sterility Non-sterile
Viscosity Data not available.
Density 0.9970 g/mL (at 20°C)
Quantity 1 L
Specific Gravity 0.99707 (at 20°C)
Physical Form Liquid
Shipping Condition Ambient Temperature
Product Line Vion Histology
For Use With (Application) Histology
Chemical Name or Material Deionized Water
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Category Not a hazardous substance.
ShelfLife 3 Years
DOTInformation Not regulated.
EINECSNumber 231-791-2
RTECSNumber EU9800000
Recommended Storage Store in a cool area away from water-reactive or dangerous when wet substances.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.