Learn More
Crescent Chemical Co Inc WATER DEMIN. STERILE
SDP
Supplier:  Crescent Chemical Co Inc SE39800.01
Encompass_Preferred
Water demin., sterile molecular biology grade DNase/RNase not detected. Autoclaved. Size - 500ML Storage Conditions - +15 °C TO +30 °C Catalog Number - 39800.01
Catalog No. 50-248-489