Learn More
Crescent Chemical Co Inc WATER DEPC (0.1%) STERILE
SDP
Catalog No. 50248485
Encompass_Preferred
Water DEPC (0.1 %) treated, sterile molecular biology grade DNase/RNase not detected. Autoclaved. Size - 500ML Storage Conditions - +15 °C TO +30 °C Catalog Number - 39798.03
Catalog No. 50-248-485 Supplier Crescent Chemical Co Inc Supplier No. SE39798.03
Water DEPC (0.1 %) treated, sterile molecular biology grade DNase/RNase not detected. Autoclaved. Size - 500ML Storage Conditions - +15 °C TO +30 °C Catalog Number - 39798.03