Learn More
Growcells WATER DEPC TREATED 125ML PK4
Catalog No. 501034784
Encompass_Preferred
Molecular Biology Grade/Nuclease Free Water; 0.001% DEPC treated, 18.2 megOhm, 0.1um sterile filtered and autoclaved in TriForest polycarbonate bottles.
Catalog No. 50-103-4784 Supplier Growcells Supplier No. UPW01254
Molecular Biology Grade/Nuclease Free Water; 0.001% DEPC treated, 18.2 megOhm, 0.1um sterile filtered and autoclaved in TriForest polycarbonate bottles.