Learn More
Growcells WATER DEPC TREATED 1L CS12
Catalog No. 501034788
Encompass_Preferred
Molecular Biology Grade/Nuclease Free Water; 0.001% DEPC treated, 18.2 megOhm, 0.1um sterile filtered and autoclaved in TriForest polycarbonate bottles.
Catalog No. 50-103-4788 Supplier Growcells Supplier No. UPW100012
Molecular Biology Grade/Nuclease Free Water; 0.001% DEPC treated, 18.2 megOhm, 0.1um sterile filtered and autoclaved in TriForest polycarbonate bottles.